Learn More
Echelon Biosciences Research Labs PHOSPHATIDYLSERINE (PS) BEADS 0.1 mL
SDP
Catalog No. NC1152500
Encompass
Phosphatidylserine Beads are composed of agarose with 10 nanomoles of bound phosphatidylserine (PS) per 1ml of beads, enough for several protein binding experiments. Each 1 mL order of Phosphatidylserine Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. NC1152500 Supplier Echelon Biosciences Research Labs Supplier No. PB0PS100UL
Phosphatidylserine Beads are composed of agarose with 10 nanomoles of bound phosphatidylserine (PS) per 1ml of beads, enough for several protein binding experiments. Each 1 mL order of Phosphatidylserine Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable