Learn More
Cayman Chemical Phosphatidylserine (soy) (sodium salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502933031
Encompass_Preferred
A mixture of soy phosphatidylserines; comprises 2-10% of total phospholipids in mammals and is enriched in the brain; found mainly on the inner leaflet of the cell membrane; synthesized in the ER and mitochondria; is a precursor to phosphatidylcholine and phosphatidylethanolamine; cofactor for various signaling pathways; externalized during apoptosis as a signal for phagocytes to engulf the cell
Catalog No. 50-293-3031 Supplier Cayman Chemical Supplier No. 2584710 MG
A mixture of soy phosphatidylserines; comprises 2-10% of total phospholipids in mammals and is enriched in the brain; found mainly on the inner leaflet of the cell membrane; synthesized in the ER and mitochondria; is a precursor to phosphatidylcholine and phosphatidylethanolamine; cofactor for various signaling pathways; externalized during apoptosis as a signal for phagocytes to engulf the cell