Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphazene Base P1-T-Bu

Catalog No. 7940825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-408-25ML 1 mL
79-408-5ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 79-408-25ML Supplier Sigma Aldrich Supplier No. 7940825ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphazene Bases: a Family of Extremely Strong, Non-Ionic, Non-Charged Nitrogen-BasesSterically hindered, neutral nitrogen base with similar basicity as BEMP (Cat. No. 20025); Base for dehydrating carbamate anions to isocyanates with POCl3 or P4O10

Specifications

CAS 111324-03-9
Molecular Formula C14H39N7P2
Linear Formula C14H39N7P2
MDL Number MFCD01863108
Quantity 1 mL
Molecular Weight (g/mol) 367.45 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.