Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphazene Base P2-T-Bu Solution

Catalog No. 7941625ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-416-25ML 25 mL
79-416-1ML 1 mL
79-416-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 79-416-25ML Supplier Sigma Aldrich Supplier No. 7941625ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphazene Bases: a Family of Extremely Strong, Non-Ionic, Non-Charged Nitrogen-BasesExtremely strong and hindered neutral nitrogen base; it is about 109 times more basic than DBU [pKa (DMSO) 21.45]. Stable to hydrolysis and unaffected by alkylating agents; Highly selective base; Deprotonation of phosphines

Specifications

CAS 111324-03-9
Molecular Formula C14H39N7P2
Linear Formula C14H39N7P2
Quantity 25 mL
MDL Number MFCD01863108
Synonym 1-tert-Butyl-2,2,4,4,4-pentakis(dimethylamino)-2lambda5,4lambda5-catenadi(phosphazene)
Molecular Weight (g/mol) 367.45 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.