Learn More
AdipoGen Phosphazenium P2-BF4
SDP
Catalog No. 502560807
Encompass_Preferred
Chemical. CAS 137334-98-6. Formula C12H36BF4N7P2. MW 427.21. Synthetic A phase transfer catalyst directing enolate alkylation towards O-substitution. Starting material for preparing the corresponding fluoride, a very reactive naked fluoride, with KF.
Catalog No. 50-256-0807 Supplier AdipoGen Supplier No. CDXH0011G001
Chemical. CAS 137334-98-6. Formula C12H36BF4N7P2. MW 427.21. Synthetic A phase transfer catalyst directing enolate alkylation towards O-substitution. Starting material for preparing the corresponding fluoride, a very reactive naked fluoride, with KF.