Learn More
Cayman Chemical PhosphIn-bIotIn 500ug
SDP
Catalog No. 502953616
Encompass_Preferred
A labeling reagent that selectively reacts with azido groups on modified proteins through the Staudinger ligation reaction; can be used in common avidin-based biochemical techniques in whole cells or by blotting experiments following SDS-PAGE; has been used successfully in conjunction with Daz-1 or Daz-2 to label and detect sulfenic acid sites in proteins
Catalog No. 50-295-3616 Supplier Cayman Chemical Supplier No. 13581500 UG
A labeling reagent that selectively reacts with azido groups on modified proteins through the Staudinger ligation reaction; can be used in common avidin-based biochemical techniques in whole cells or by blotting experiments following SDS-PAGE; has been used successfully in conjunction with Daz-1 or Daz-2 to label and detect sulfenic acid sites in proteins