Learn More
Cayman Chemical Phosphine-biotin, Purity: ≥95%, CAS: 608514-42-7, 5 mg
SDP
Catalog No. 502955049
Encompass_Preferred
A labeling reagent that selectively reacts with azido groups on modified proteins through the Staudinger ligation reaction; can be used in common avidin-based biochemical techniques in whole cells or by blotting experiments following SDS-PAGE; has been used successfully in conjunction with Daz-1 or Daz-2 to label and detect sulfenic acid sites in proteins
Catalog No. 50-295-5049 Supplier Cayman Chemical Supplier No. 135815 MG
A labeling reagent that selectively reacts with azido groups on modified proteins through the Staudinger ligation reaction; can be used in common avidin-based biochemical techniques in whole cells or by blotting experiments following SDS-PAGE; has been used successfully in conjunction with Daz-1 or Daz-2 to label and detect sulfenic acid sites in proteins