Learn More
Medchemexpress LLC Phosphine, bis(tricyclo[3.3.1.13,7]dec-1-yl)- | 131211-27-3 | MFCD06658117 | 97.0% | 302.43 | 10 G
SDP
Catalog No. 5000395698
Encompass_Preferred
Di(adamantan-1-yl)phosphine, also known as Di-1-adamantylphosphine, is a biological molecule intended for research use only. This solid appears white to off-white and is crucial for various biochemical assays.
  • Store at 4°C, protected from light, and under nitrogen.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light and under nitrogen.
  • For research use only.
Catalog No. 50-003-95698 Supplier Medchemexpress LLC Supplier No. HYW00854810G
Di(adamantan-1-yl)phosphine, also known as Di-1-adamantylphosphine, is a biological molecule intended for research use only. This solid appears white to off-white and is crucial for various biochemical assays.
  • Store at 4°C, protected from light, and under nitrogen.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light and under nitrogen.
  • For research use only.