Learn More
Strem, An Ascensus Company CAS# 15104-46-8. 25g. 1,2-Bis(dimethoxyphosphoryl)benzene, 99%. MFCD00142461
SDP
Supplier:  Strem, An Ascensus Company 15008925G
Encompass
CAS# 15104-46-8. 25g. 1,2-Bis(dimethoxyphosphoryl)benzene, 99%. MFCD00142461. Molecular Weight: 294.18. Molecular Formula: C6H4[P(O)(OCH3)2]2. Color/form: white xtl. Strem# 15-0089. http://www.strem.com/catalog/v/15-0089/
Catalog No. 50-204-5231