Learn More
Medchemexpress LLC Phosphine, diphenyl-1-pyrenyl- | 110231-30-6 | 95.0% | 386.42 | C28H19P | 50 MG
SDP
Catalog No. 5000228710
Encompass_Preferred
Diphenyl-1-pyrenylphosphine (DPPP) is a phosphine-based fluorogenic probe used to detect peroxides and lipid peroxidation in biological samples. It exhibits phototriggered aggregation-induced emission/aggregation-caused quenching behavior and is supplied as a solid reagent for research applications.
  • Fluorogenic probe that reacts with peroxides to yield a fluorescent product.
  • Sensitive detection of lipid peroxidation in cells and tissues.
  • Exhibits phototriggered AIE/ACQ transition for enhanced fluorescence control.
  • Off-white to light yellow solid, suitable for laboratory handling and storage.
  • Store protected from light; stable under recommended cold storage conditions.
Catalog No. 50-002-28710 Supplier Medchemexpress LLC Supplier No. HY11815950MG
Diphenyl-1-pyrenylphosphine (DPPP) is a phosphine-based fluorogenic probe used to detect peroxides and lipid peroxidation in biological samples. It exhibits phototriggered aggregation-induced emission/aggregation-caused quenching behavior and is supplied as a solid reagent for research applications.
  • Fluorogenic probe that reacts with peroxides to yield a fluorescent product.
  • Sensitive detection of lipid peroxidation in cells and tissues.
  • Exhibits phototriggered AIE/ACQ transition for enhanced fluorescence control.
  • Off-white to light yellow solid, suitable for laboratory handling and storage.
  • Store protected from light; stable under recommended cold storage conditions.