Learn More
Medchemexpress LLC Phosphine, diphenyl-1-pyrenyl- | 110231-30-6 | MFCD00278755 | 95.0% | 386.42 | C28H19P | 10 MG
SDP
Catalog No. 5000226939
Encompass_Preferred
Diphenyl-1-pyrenylphosphine (DPPP) is a fluorogenic peroxide-reactive probe used to detect hydroperoxides and reactive oxygen species in biochemical and cellular assays. Upon reaction with hydroperoxides it converts to a fluorescent phosphine oxide, enabling sensitive measurement of lipid peroxidation and oxidative stress.
  • Fluorogenic hydroperoxide probe enabling sensitive detection.
  • Reacts stoichiometrically with hydroperoxides to yield a fluorescent product.
  • Suitable for assays of lipid peroxidation and oxidative stress in biological samples.
  • Supplied as a solid with typical purity around 95.0%.
  • Molecular formula C28H19P; molecular weight 386.42.
Catalog No. 50-002-26939 Supplier Medchemexpress LLC Supplier No. HY11815910MG
Diphenyl-1-pyrenylphosphine (DPPP) is a fluorogenic peroxide-reactive probe used to detect hydroperoxides and reactive oxygen species in biochemical and cellular assays. Upon reaction with hydroperoxides it converts to a fluorescent phosphine oxide, enabling sensitive measurement of lipid peroxidation and oxidative stress.
  • Fluorogenic hydroperoxide probe enabling sensitive detection.
  • Reacts stoichiometrically with hydroperoxides to yield a fluorescent product.
  • Suitable for assays of lipid peroxidation and oxidative stress in biological samples.
  • Supplied as a solid with typical purity around 95.0%.
  • Molecular formula C28H19P; molecular weight 386.42.