Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphine, diphenyl-1-pyrenyl- | 110231-30-6 | MFCD00278755 | 95.0% | 386.42 | C28H19P | 5 MG
SDP

Catalog No. 5000226940
Encompass_Preferred

Diphenyl-1-pyrenylphosphine (DPPP) is a fluorogenic peroxide-reactive probe used to detect hydroperoxides and study photophysical behavior. It exhibits phototriggered aggregation-induced emission (AIE) / aggregation-caused quenching (ACQ) transitions and notable third-order nonlinear optical responses. Typically supplied as a solid in small milligram quantities for research use.

  • Fluorogenic probe for peroxide and ROS detection.
  • Photophysical behavior: AIE/ACQ transitions and third-order nonlinear optical response.
  • Purity 95.0%.
  • Molecular weight 386.42 Da.
  • Chemical formula C28H19P.
  • Supplied as a solid powder in milligram quantities.

Catalog No. 50-002-26940 Supplier Medchemexpress LLC Supplier No. HY1181595MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenyl-1-pyrenylphosphine (DPPP) is a fluorogenic peroxide-reactive probe used to detect hydroperoxides and study photophysical behavior. It exhibits phototriggered aggregation-induced emission (AIE) / aggregation-caused quenching (ACQ) transitions and notable third-order nonlinear optical responses. Typically supplied as a solid in small milligram quantities for research use.

  • Fluorogenic probe for peroxide and ROS detection.
  • Photophysical behavior: AIE/ACQ transitions and third-order nonlinear optical response.
  • Purity 95.0%.
  • Molecular weight 386.42 Da.
  • Chemical formula C28H19P.
  • Supplied as a solid powder in milligram quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.