Learn More
Strem, An Ascensus Company CAS 205495-66-5. 2g. (1S2S)-(-)-12-Diaminocyclohexane-NN-bis(2-diphenylphosphino-1-naphthoyl) min. 94% (SS)-DACH-Naphthyl Trost Ligand. MFCD02684552
SDP
Supplier:  Strem, An Ascensus Company 1509642G
Encompass
CAS 205495-66-5. 2g. (1S2S)-(-)-12-Diaminocyclohexane-NN-bis(2-diphenylphosphino-1-naphthoyl) min. 94% (SS)-DACH-Naphthyl Trost Ligand. MFCD02684552. Molecular Weight 790.88. Molecular Formula C52H44N2O2P2. Color/form off-white pwdr. Strem 15-0964. http//www.strem.com/catalog/v/15-0964/
Catalog No. 50-211-0743