Learn More
Gold Biotechnology Inc Phosphinothricin 1 g
SDP
Catalog No. 501532821
Encompass_Preferred
Phosphinothricin is a broad-spectrum herbicide that acts as a competitive inhibitor of glutamine synthetase.
Catalog No. 50-153-2821 Supplier Gold Biotechnology Inc Supplier No. P1651
Phosphinothricin is a broad-spectrum herbicide that acts as a competitive inhibitor of glutamine synthetase.