Learn More
Cell Signaling Technology Phospho-4E-BP1 (Ser65) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503133343
Encompass
The Phospho-4E-BP1 (Ser65) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling, including the use of fluorophores, lanthanides, biotin, or beads. This pair consists of capture and detection antibodies that bind to non-overlapping epitopes, supporting various conjugation-reliant platforms.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)), BSA and azide free.
  • Validated for human species reactivity.
  • Optimal dilutions and concentrations should be determined by the end user.
Catalog No. 50-313-3343 Supplier Cell Signaling Technology Supplier No. 99459S
The Phospho-4E-BP1 (Ser65) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling, including the use of fluorophores, lanthanides, biotin, or beads. This pair consists of capture and detection antibodies that bind to non-overlapping epitopes, supporting various conjugation-reliant platforms.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)), BSA and azide free.
  • Validated for human species reactivity.
  • Optimal dilutions and concentrations should be determined by the end user.