Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-AKT(S473) Antibody (YA5759) | 56 kDa | 10 UL
SDP

Catalog No. 5000447055
Encompass_Preferred

Phospho-AKT(S473) Antibody (YA5759) is a mouse-derived, non-conjugated monoclonal antibody specifically designed to target Phospho-AKT(S473). This antibody is supplied in a liquid formulation containing PBS, 50% glycerol, 0.5% BSA, and 0.02% sodium azide. It is purified through affinity chromatography, ensuring high specificity for research applications.

  • For research use only
  • Monoclonal antibody, mouse-derived
  • Targets Phospho-AKT(S473)
  • Supplied in PBS with 50% glycerol, 0.5% BSA, 0.02% sodium azide
  • Appearance: liquid
  • Storage at -20°C for up to 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Catalog No. 50-004-47055 Supplier Medchemexpress LLC Supplier No. HYP8606710UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-AKT(S473) Antibody (YA5759) is a mouse-derived, non-conjugated monoclonal antibody specifically designed to target Phospho-AKT(S473). This antibody is supplied in a liquid formulation containing PBS, 50% glycerol, 0.5% BSA, and 0.02% sodium azide. It is purified through affinity chromatography, ensuring high specificity for research applications.

  • For research use only
  • Monoclonal antibody, mouse-derived
  • Targets Phospho-AKT(S473)
  • Supplied in PBS with 50% glycerol, 0.5% BSA, 0.02% sodium azide
  • Appearance: liquid
  • Storage at -20°C for up to 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.