Learn More
Medchemexpress LLC Phospho-AKT(S473) Antibody (YA5759) | 56 kDa | 100 UL
SDP
Catalog No. 5000447166
Encompass_Preferred
Phospho-AKT(S473) Antibody (YA5759) is a mouse-derived, non-conjugated monoclonal antibody targeting Phospho-AKT(S473). AKT protein is crucial in regulating cellular processes like metabolism, proliferation, cell survival, growth, and angiogenesis, functioning through serine and/or threonine phosphorylation of numerous downstream substrates.
  • Supplied in PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipping with blue ice
  • Suitable for WB, IHC-P, and ICC/IF applications
Catalog No. 50-004-47166 Supplier Medchemexpress LLC Supplier No. HYP86067100UL
Phospho-AKT(S473) Antibody (YA5759) is a mouse-derived, non-conjugated monoclonal antibody targeting Phospho-AKT(S473). AKT protein is crucial in regulating cellular processes like metabolism, proliferation, cell survival, growth, and angiogenesis, functioning through serine and/or threonine phosphorylation of numerous downstream substrates.
  • Supplied in PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipping with blue ice
  • Suitable for WB, IHC-P, and ICC/IF applications