Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-AKT(S473) antibody (YA5759) | 56 kDa | 50 UL
SDP

Catalog No. 5000449115
Encompass_Preferred

Phospho-AKT(S473) antibody (YA5759) is a mouse-derived, non-conjugated monoclonal antibody targeting Phospho-AKT(S473). The AKT protein, a member of the AKT kinase family, is pivotal in regulating cellular processes including metabolism, proliferation, survival, growth, and angiogenesis. It functions by phosphorylating numerous downstream substrates, with AKT1 involved in modulating over 100 candidates, orchestrating glucose uptake and storage, protein synthesis, and lipid metabolism. This antibody is a key tool for research into these complex cellular pathways.

  • Formulation: supplied in PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide.
  • Purity: affinity chromatography.
  • Storage and stability: stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
  • Appearance: liquid.
  • Applications: WB (1:1000-2000), IHC-P (1:100-200), ICC/IF (1:50-200).
  • Shipping: shipped with blue ice.

Catalog No. 50-004-49115 Supplier Medchemexpress LLC Supplier No. HYP8606750UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-AKT(S473) antibody (YA5759) is a mouse-derived, non-conjugated monoclonal antibody targeting Phospho-AKT(S473). The AKT protein, a member of the AKT kinase family, is pivotal in regulating cellular processes including metabolism, proliferation, survival, growth, and angiogenesis. It functions by phosphorylating numerous downstream substrates, with AKT1 involved in modulating over 100 candidates, orchestrating glucose uptake and storage, protein synthesis, and lipid metabolism. This antibody is a key tool for research into these complex cellular pathways.

  • Formulation: supplied in PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide.
  • Purity: affinity chromatography.
  • Storage and stability: stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
  • Appearance: liquid.
  • Applications: WB (1:1000-2000), IHC-P (1:100-200), ICC/IF (1:50-200).
  • Shipping: shipped with blue ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.