Learn More
BIOHIPPO Phospho-AKT1 Ser473 rAb W-R33
SDP
Catalog No. 5000975456
Encompass
U-Blot Phospho-AKT1 (Ser473) Rabbit mAb is a Rabbit-derived recombinant monoclonal antibody (clone W-R33) that specifically recognizes Phospho-AKT1 (Ser473) a phosphorylation modification at Ser473 It shows reactivity with Human Rat Validated applications include WB Recommended dilutions WB 11000-12000 Purified by protein a AKT1 is one of 3 closely related serine/threonine-protein kinases (AKT1 AKT2 and AKT3) called the AKT kinase which regulate many processes including metabolism proliferation cell survival growth and angiogenesis AKT has an important role in the regulation of NF-kappa-B-dependent gene transcription and positively regulates the activity of CREB1 (cyclic AMP (cAMP)-response element binding protein) Supplied as 100 uL
Catalog No. 50-009-75456 Supplier BIOHIPPO Supplier No. BHA21821342
U-Blot Phospho-AKT1 (Ser473) Rabbit mAb is a Rabbit-derived recombinant monoclonal antibody (clone W-R33) that specifically recognizes Phospho-AKT1 (Ser473) a phosphorylation modification at Ser473 It shows reactivity with Human Rat Validated applications include WB Recommended dilutions WB 11000-12000 Purified by protein a AKT1 is one of 3 closely related serine/threonine-protein kinases (AKT1 AKT2 and AKT3) called the AKT kinase which regulate many processes including metabolism proliferation cell survival growth and angiogenesis AKT has an important role in the regulation of NF-kappa-B-dependent gene transcription and positively regulates the activity of CREB1 (cyclic AMP (cAMP)-response element binding protein) Supplied as 100 uL