Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-AMPK alpha 1 (Ser496) Antibody | 100 UL
SDP

Catalog No. 5000442659
Encompass_Preferred

This rabbit-derived, non-conjugated IgG polyclonal antibody specifically targets phosphorylated AMPK alpha 1 at Ser496. Intended for research use only, it serves as a valuable tool for detecting modification-specific phosphorylation, crucial for studies in cellular energy metabolism.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human
  • Molecular weight: 64 kDa (predicted and observed)
  • Immunogen: Synthetic phosphopeptide (Ser496 of Human AMPKα1)
  • Applications: Western blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Purity: Affinity purified
  • Formulation: Supplied in 1*PBS (pH 7.3), 50% glycerol, 0.5% BSA, 0.02% sodium azide
  • Storage: Store at -20°C for 1 year, avoid freeze/thaw cycles
  • Shipping: With blue ice
  • Detects phosphorylated protein at Ser496
  • Supports research in cellular energy metabolism

Catalog No. 50-004-42659 Supplier Medchemexpress LLC Supplier No. HYP80790100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit-derived, non-conjugated IgG polyclonal antibody specifically targets phosphorylated AMPK alpha 1 at Ser496. Intended for research use only, it serves as a valuable tool for detecting modification-specific phosphorylation, crucial for studies in cellular energy metabolism.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human
  • Molecular weight: 64 kDa (predicted and observed)
  • Immunogen: Synthetic phosphopeptide (Ser496 of Human AMPKα1)
  • Applications: Western blot (WB), Immunohistochemistry (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Purity: Affinity purified
  • Formulation: Supplied in 1*PBS (pH 7.3), 50% glycerol, 0.5% BSA, 0.02% sodium azide
  • Storage: Store at -20°C for 1 year, avoid freeze/thaw cycles
  • Shipping: With blue ice
  • Detects phosphorylated protein at Ser496
  • Supports research in cellular energy metabolism

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.