Learn More
Cell Signaling Technology Matched Antibody Pair, Phospho-AMPKα, Human and Mouse, 1 Kit
SDP
Catalog No. 503131758
Encompass
The Phospho-AMPKα (Thr172) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This kit is suitable for use in platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Contains capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).
  • BSA and azide free formulation.
  • Reactive in human and mouse models.
  • Includes AMPK alpha (23A3) Rabbit Monoclonal Antibody and Phospho-AMPK alpha (Thr172) (A9S3O) Mouse Monoclonal Antibody.
Catalog No. 50-313-1758 Supplier Cell Signaling Technology Supplier No. 16360S
The Phospho-AMPKα (Thr172) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This kit is suitable for use in platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Contains capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).
  • BSA and azide free formulation.
  • Reactive in human and mouse models.
  • Includes AMPK alpha (23A3) Rabbit Monoclonal Antibody and Phospho-AMPK alpha (Thr172) (A9S3O) Mouse Monoclonal Antibody.