Learn More
Cell Signaling Technology Phospho-Axl (Tyr702) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131872
Encompass
The Phospho-Axl (Tyr702) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, and beads. This kit features capture and detection antibodies that bind to non-overlapping epitopes, supporting use in platforms like MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Ideal for high-throughput platforms requiring custom antibody conjugation.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)).
  • Formulated BSA and azide free.
  • Consists of capture and detection antibodies that bind to non-overlapping epitopes.
  • Validated for human reactivity.
  • Store at -20°C.
Catalog No. 50-313-1872 Supplier Cell Signaling Technology Supplier No. 28837S
The Phospho-Axl (Tyr702) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, and beads. This kit features capture and detection antibodies that bind to non-overlapping epitopes, supporting use in platforms like MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Ideal for high-throughput platforms requiring custom antibody conjugation.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)).
  • Formulated BSA and azide free.
  • Consists of capture and detection antibodies that bind to non-overlapping epitopes.
  • Validated for human reactivity.
  • Store at -20°C.