Learn More
Cell Signaling Technology Phospho-Bad (Ser112) Matched Antibody Pair, Unconjugated, 1 Kit
SDP
Catalog No. 503134206
Encompass
The Phospho-Bad (Ser112) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom or specialized labeling, including biotin, fluorophores, lanthanides, and beads. This kit is compatible with platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in BSA- and azide-free formulation.
  • Validated for reactivity in human, mouse, rat, and monkey samples.
  • Suitable for applications including ELISA and related immunoassay technologies.
Catalog No. 50-313-4206 Supplier Cell Signaling Technology Supplier No. 61300S
The Phospho-Bad (Ser112) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom or specialized labeling, including biotin, fluorophores, lanthanides, and beads. This kit is compatible with platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in BSA- and azide-free formulation.
  • Validated for reactivity in human, mouse, rat, and monkey samples.
  • Suitable for applications including ELISA and related immunoassay technologies.