Learn More
Cell Signaling Technology Phospho-c-Fos (Ser32) Matched Antibody Pair, Unconjugated, Human, Mouse, 1 Kit
SDP
Catalog No. 503131967
Encompass
The Phospho-c-Fos (Ser32) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and is ideal for applications requiring conjugated antibody pairs, such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes c-Fos (E2I7R) rabbit monoclonal antibody and Phospho-c-Fos (Ser32) (D82C12) rabbit monoclonal antibody
  • Supplied in BSA and azide-free formulation
  • Compatible with diverse labeling technologies including fluorophores, lanthanides, biotin, and beads
  • Validated for human and mouse species reactivity
  • Optimized for ELISA and ELISA-like assays
Catalog No. 50-313-1967 Supplier Cell Signaling Technology Supplier No. 37524S
The Phospho-c-Fos (Ser32) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and is ideal for applications requiring conjugated antibody pairs, such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes c-Fos (E2I7R) rabbit monoclonal antibody and Phospho-c-Fos (Ser32) (D82C12) rabbit monoclonal antibody
  • Supplied in BSA and azide-free formulation
  • Compatible with diverse labeling technologies including fluorophores, lanthanides, biotin, and beads
  • Validated for human and mouse species reactivity
  • Optimized for ELISA and ELISA-like assays