Learn More
Cell Signaling Technology Phospho-c-Kit (panTyr) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131836
Encompass
The Phospho-c-Kit (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads. This antibody pair is compatible with various platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Optimized for high-throughput platforms
  • Compatible with custom labeling and conjugation
  • Supplied in BSA and azide-free formulation
  • Validated for human species reactivity
  • Detection of phosphorylated c-Kit in a range of assay formats
Catalog No. 50-313-1836 Supplier Cell Signaling Technology Supplier No. 26936S
The Phospho-c-Kit (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads. This antibody pair is compatible with various platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Optimized for high-throughput platforms
  • Compatible with custom labeling and conjugation
  • Supplied in BSA and azide-free formulation
  • Validated for human species reactivity
  • Detection of phosphorylated c-Kit in a range of assay formats