Learn More
Cell Signaling Technology Phospho-c-Kit (Tyr703) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132342
Encompass
The Phospho-c-Kit (Tyr703) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and is supplied in a BSA and azide-free formulation.
  • Ideal for conjugation with fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Consists of capture and detection antibodies binding to non-overlapping epitopes.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).
  • BSA and azide free.
  • Recommended to be aliquoted into single-use vials to avoid multiple freeze/thaw cycles.
Catalog No. 50-313-2342 Supplier Cell Signaling Technology Supplier No. 33237S
The Phospho-c-Kit (Tyr703) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and is supplied in a BSA and azide-free formulation.
  • Ideal for conjugation with fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Consists of capture and detection antibodies binding to non-overlapping epitopes.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).
  • BSA and azide free.
  • Recommended to be aliquoted into single-use vials to avoid multiple freeze/thaw cycles.