Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-c-Myc(Ser62) Antibody (YA211) | Predicted band size: 49 kDa; Observed band size: 57 kDa | 50 UL
SDP

Catalog No. 5000396033
Encompass_Preferred

The Phospho-c-Myc(Ser62) antibody is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-c-Myc(Ser62). It is suitable for various laboratory research applications, including Western Blot, immunocytochemistry/immunofluorescence, immunohistochemistry-paraffin, and immunoprecipitation.

  • Host: rabbit
  • Reactivity: human
  • Isotype: IgG monoclonal antibody
  • Conjugation: non-conjugated
  • Applications: WB, ICC/IF, IHC-P, IP
  • Formulation: supplied in 1*TBS (pH7.4), 0.05% BSA, 40% glycerol, and 0.05% sodium azide
  • Storage: store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Shipping: shipped with blue ice

Catalog No. 50-003-96033 Supplier Medchemexpress LLC Supplier No. HYP8051150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-c-Myc(Ser62) antibody is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-c-Myc(Ser62). It is suitable for various laboratory research applications, including Western Blot, immunocytochemistry/immunofluorescence, immunohistochemistry-paraffin, and immunoprecipitation.

  • Host: rabbit
  • Reactivity: human
  • Isotype: IgG monoclonal antibody
  • Conjugation: non-conjugated
  • Applications: WB, ICC/IF, IHC-P, IP
  • Formulation: supplied in 1*TBS (pH7.4), 0.05% BSA, 40% glycerol, and 0.05% sodium azide
  • Storage: store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Shipping: shipped with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.