Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-CD18 (Ser745) Antibody (YA220) | 10 UL
SDP

Catalog No. 5000442865
Encompass_Preferred

The Phospho-CD18 (Ser745) Antibody (YA220) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically designed to target Phospho-CD18 (Ser745). This antibody is suitable for use in research applications such as Western Blot (WB) and Immunoprecipitation (IP), and has been validated for reactivity across human, mouse, and rat species.

  • Rabbit-derived monoclonal antibody
  • Targets phospho-CD18 (Ser745)
  • Non-conjugated IgG type
  • Predicted and observed molecular weight is 85 kDa
  • Purity is affinity purified
  • Reactive in human, mouse, and rat samples
  • Applications include Western Blot and Immunoprecipitation
  • Supplied in PBS, pH 7.4
  • Storage at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Catalog No. 50-004-42865 Supplier Medchemexpress LLC Supplier No. HYP8079510UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-CD18 (Ser745) Antibody (YA220) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically designed to target Phospho-CD18 (Ser745). This antibody is suitable for use in research applications such as Western Blot (WB) and Immunoprecipitation (IP), and has been validated for reactivity across human, mouse, and rat species.

  • Rabbit-derived monoclonal antibody
  • Targets phospho-CD18 (Ser745)
  • Non-conjugated IgG type
  • Predicted and observed molecular weight is 85 kDa
  • Purity is affinity purified
  • Reactive in human, mouse, and rat samples
  • Applications include Western Blot and Immunoprecipitation
  • Supplied in PBS, pH 7.4
  • Storage at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.