Learn More
Cell Signaling Technology Polyclonal Antibody, Phospho-Cdc25C (Thr48), Rabbit, Human, 20 μL
SDP
Catalog No. 502729833
Encompass
This phospho-specific polyclonal antibody detects endogenous levels of cdc25C only when phosphorylated at Thr48. The antibody is developed by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding Thr48 of human cdc25C and is purified via protein A and peptide affinity chromatography. This reagent is suitable for investigation of cdc25C regulation in human cell cycle studies.
  • Validated for western blotting applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol
  • Target reactivity specific to human
  • Endogenous sensitivity
Catalog No. 50-272-9833 Supplier Cell Signaling Technology Supplier No. 9527T
This phospho-specific polyclonal antibody detects endogenous levels of cdc25C only when phosphorylated at Thr48. The antibody is developed by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding Thr48 of human cdc25C and is purified via protein A and peptide affinity chromatography. This reagent is suitable for investigation of cdc25C regulation in human cell cycle studies.
  • Validated for western blotting applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol
  • Target reactivity specific to human
  • Endogenous sensitivity