Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-CDK1/2/3 (Thr14) Antibody (YA218) | 100 UL
SDP

Catalog No. 5000442664
Encompass_Preferred

The Phospho-CDK1/2/3 (Thr14) Antibody (YA218) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically designed to target Phospho-CDK1/2/3 (Thr14). This antibody is for research use only, aiding in the study of cell cycle regulation. CDK1/2/3 are pivotal kinases involved in controlling the eukaryotic cell cycle, modulating centrosome dynamics and mitotic onset, and influencing G2-M transitions.

  • Rabbit-derived, monoclonal IgG antibody
  • Non-conjugated
  • Targets Phospho-CDK1/2/3 (Thr14)
  • Reactivity with human and hamster species
  • Molecular weight: 34 kDa
  • Application for Western Blot (WB) at 1:500-1:1000 dilution
  • Application for Immunoprecipitation (IP) at 1:20 dilution
  • Formulated in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, and 0.05% BSA

Catalog No. 50-004-42664 Supplier Medchemexpress LLC Supplier No. HYP80797100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-CDK1/2/3 (Thr14) Antibody (YA218) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically designed to target Phospho-CDK1/2/3 (Thr14). This antibody is for research use only, aiding in the study of cell cycle regulation. CDK1/2/3 are pivotal kinases involved in controlling the eukaryotic cell cycle, modulating centrosome dynamics and mitotic onset, and influencing G2-M transitions.

  • Rabbit-derived, monoclonal IgG antibody
  • Non-conjugated
  • Targets Phospho-CDK1/2/3 (Thr14)
  • Reactivity with human and hamster species
  • Molecular weight: 34 kDa
  • Application for Western Blot (WB) at 1:500-1:1000 dilution
  • Application for Immunoprecipitation (IP) at 1:20 dilution
  • Formulated in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, and 0.05% BSA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.