Learn More
Medchemexpress LLC Phospho-CDK1/2/3 (Thr14) Antibody (YA218) | 50 UL
SDP
Catalog No. 5000441957
Encompass_Preferred
Phospho-CDK1/2/3 (Thr14) Antibody (YA218) is a Rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting the phosphorylated Thr14 site of CDK1, CDK2, and CDK3. These cyclin-dependent kinases are pivotal regulators of the eukaryotic cell cycle, controlling G0-G1, G1-S, and G2-M transitions, DNA synthesis, and cell proliferation. This antibody supports cell biology research into the regulatory mechanisms and subcellular localization of these essential kinases.
  • Rabbit-derived monoclonal antibody
  • Targets phosphorylated Thr14 of CDK1, CDK2, and CDK3
  • Essential for studying cell cycle regulation
  • Aids research into DNA synthesis and cell proliferation
  • Non-conjugated IgG isotype
  • Suitable for Western Blot (WB) and Immunoprecipitation (IP) applications
  • Predicted and observed molecular weight: 34 kDa
  • Affinity purified for high specificity
  • Stable for 1 year when stored at -20°C
Catalog No. 50-004-41957 Supplier Medchemexpress LLC Supplier No. HYP8079750UL
Phospho-CDK1/2/3 (Thr14) Antibody (YA218) is a Rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting the phosphorylated Thr14 site of CDK1, CDK2, and CDK3. These cyclin-dependent kinases are pivotal regulators of the eukaryotic cell cycle, controlling G0-G1, G1-S, and G2-M transitions, DNA synthesis, and cell proliferation. This antibody supports cell biology research into the regulatory mechanisms and subcellular localization of these essential kinases.
  • Rabbit-derived monoclonal antibody
  • Targets phosphorylated Thr14 of CDK1, CDK2, and CDK3
  • Essential for studying cell cycle regulation
  • Aids research into DNA synthesis and cell proliferation
  • Non-conjugated IgG isotype
  • Suitable for Western Blot (WB) and Immunoprecipitation (IP) applications
  • Predicted and observed molecular weight: 34 kDa
  • Affinity purified for high specificity
  • Stable for 1 year when stored at -20°C