Learn More
Cell Signaling Technology Phospho-Chk1 (Ser345) Matched Antibody Pair, Human Reactivity, ELISA, 1 Kit
SDP
Catalog No. 503132216
Encompass
The Phospho-Chk1 (Ser345) Matched Antibody Pair provides a pair of capture and detection antibodies specifically developed for use in immunoassays and high-throughput platforms. This pair is supplied in a BSA and azide-free formulation, making it compatible with custom labeling methods including fluorophores, lanthanides, biotin, and beads. The antibodies are designed for use with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Compatible with platforms like MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Supplied in 1X PBS (pH 7.8)
  • BSA and azide-free formulation suitable for custom labeling
  • Validated for human species reactivity
  • For research use only; not for use in diagnostic procedures
Catalog No. 50-313-2216 Supplier Cell Signaling Technology Supplier No. 29860S
The Phospho-Chk1 (Ser345) Matched Antibody Pair provides a pair of capture and detection antibodies specifically developed for use in immunoassays and high-throughput platforms. This pair is supplied in a BSA and azide-free formulation, making it compatible with custom labeling methods including fluorophores, lanthanides, biotin, and beads. The antibodies are designed for use with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Compatible with platforms like MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Supplied in 1X PBS (pH 7.8)
  • BSA and azide-free formulation suitable for custom labeling
  • Validated for human species reactivity
  • For research use only; not for use in diagnostic procedures