Learn More
Enzo Life Sciences CREB (pSer133) monoclonal antibody (10E9) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905639100
Encompass_Preferred
Clone: 10E9. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of CREB phosphorylated at Ser133. UniProt: P16220 (human). Species Reactivity: Human, Mouse, Dog. Applications: ELISA, WB. Recommended dilutions: ELISA (0.05µg/ml). Western Blot (0.5µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-658-395