Learn More
Medchemexpress LLC Phospho-delta 1 Catenin (Ser268) Antibody (YA1019) | 100 UL
SDP
Catalog No. 5000442987
Encompass_Preferred
The Phospho-delta 1 Catenin (Ser268) Antibody (YA1019) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets Phospho-delta 1 Catenin (Ser268).
  • Derived from rabbit
  • Non-conjugated IgG monoclonal antibody
  • Specific for Phospho-delta 1 Catenin (Ser268)
  • Applicable in Western Blot (WB)
  • Reacts with human samples
  • Reacts with mouse samples
Catalog No. 50-004-42987 Supplier Medchemexpress LLC Supplier No. HYP81296A100UL
The Phospho-delta 1 Catenin (Ser268) Antibody (YA1019) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets Phospho-delta 1 Catenin (Ser268).
  • Derived from rabbit
  • Non-conjugated IgG monoclonal antibody
  • Specific for Phospho-delta 1 Catenin (Ser268)
  • Applicable in Western Blot (WB)
  • Reacts with human samples
  • Reacts with mouse samples