Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-delta 1 Catenin (Ser268) Antibody (YA1019) | 50 UL
SDP

Catalog No. 5000441337
Encompass_Preferred

This is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Phospho-delta 1 Catenin (Ser268).

  • Rabbit-derived
  • Non-conjugated IgG monoclonal antibody
  • Specifically targets Phospho-delta 1 Catenin (Ser268)
  • Application: Western blot
  • Reactivity: Human
  • Reactivity: Mouse

Catalog No. 50-004-41337 Supplier Medchemexpress LLC Supplier No. HYP81296A50UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Phospho-delta 1 Catenin (Ser268).

  • Rabbit-derived
  • Non-conjugated IgG monoclonal antibody
  • Specifically targets Phospho-delta 1 Catenin (Ser268)
  • Application: Western blot
  • Reactivity: Human
  • Reactivity: Mouse

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.