Learn More
Medchemexpress LLC Phospho-DRP1(Ser616) Antibody (YA5812) | 82 kDa | 10 UL
SDP
Catalog No. 5000448913
Encompass_Preferred
This Phospho-DRP1(Ser616) antibody (YA5812) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Phospho-DRP1(Ser616). DNM1L, also known as DRP1, is a key protein involved in mitochondrial and peroxisomal division, mediating membrane fission. This antibody can be used to detect the phosphorylated form of DRP1 at Ser616.
  • Western blot (WB) applications with dilution ratio of 1:2000-1:10000
  • Immunocytochemistry/immunofluorescence (ICC/IF) applications with dilution ratio of 1:200-1:1000
  • Enzyme-linked immunosorbent assay (ELISA) applications with dilution ratio of 1:5000-1:20000
  • Immunoprecipitation (IP) applications with dilution ratio of 1:50-1:200
Catalog No. 50-004-48913 Supplier Medchemexpress LLC Supplier No. HYP8612010UL
This Phospho-DRP1(Ser616) antibody (YA5812) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Phospho-DRP1(Ser616). DNM1L, also known as DRP1, is a key protein involved in mitochondrial and peroxisomal division, mediating membrane fission. This antibody can be used to detect the phosphorylated form of DRP1 at Ser616.
  • Western blot (WB) applications with dilution ratio of 1:2000-1:10000
  • Immunocytochemistry/immunofluorescence (ICC/IF) applications with dilution ratio of 1:200-1:1000
  • Enzyme-linked immunosorbent assay (ELISA) applications with dilution ratio of 1:5000-1:20000
  • Immunoprecipitation (IP) applications with dilution ratio of 1:50-1:200