Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phospho(enol)pyruvic acid cyclohexylammonium salt

Catalog No. P36375G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P36375G 5 g
P36371G 1 g
P3637500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P36375G Supplier Sigma Aldrich Supplier No. P36375G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phospho(enol)pyruvic acid (pep) is involved in glycolysis and gluconeogenesis
  • In glycolysis, pep is metabolized by pyruvate kinase to yield pyruvate
  • In plants, pep is involved in the formation of aromatic amino acids as well as in the carbon fixation pa

Specifications

CAS 10526-80-4
Molecular Formula C3H4O6P -+ C6H14N
Linear Formula C3H4O6P -+ C6H14N
MDL Number MFCD00036375
Quantity 5 g
Synonym 2-(Phosphonooxy)-2-propenoic acid monopotassium salt
Molecular Weight (g/mol) 267.22
Percent Purity ≥97% (enzymatic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.