Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phospho(enol)pyruvic acid monopotassium salt

Catalog No. 860077250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-007-7250MG 250 mg
86-007-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-007-7250MG Supplier Sigma Aldrich Supplier No. 860077250MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phospho(enol)pyruvic acid monopotassium salt

Specifications

CAS 4265-07-0
Melting Point 175°C (lit.)
Molecular Formula C3H4KO6P
Linear Formula KOP(O)(OH)OC(=CH2)CO2H
MDL Number MFCD00044476
Quantity 250 mg
Synonym mono-Potassium phosphoenolpyruvate; PEP monopotassium salt; PEP-K; Phospho(enol)pyruvate potassium salt
Molecular Weight (g/mol) 206.13
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.