Learn More
Abcam Phospho-eNOS (S1177) and Total eNOS ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2797791X96TESTS
Encompass_Preferred
Phospho-eNOS (S1177) and Total eNOS ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-eNOS (S1177) and Total eNOS in Human in Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2200317