Learn More
ABclonal Technology Phospho-ErbB4/HER4-Y1284
SDP
Catalog No. 5000013715
Encompass_Preferred
This gene is a member of the Tyr protein kinase family and the epidermal growth factor receptor subfamily It encodes a single-pass type I membrane protein with multiple cysteine rich domains a transmembrane domain a tyrosine kinase domain a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif The protein binds to and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment Mutations in this gene have been associated with cancer Alternatively spliced variants which encode different protein isoforms have been described however not all variants have been fully characterized
Catalog No. 50-000-13715 Supplier ABclonal Technology Supplier No. AP1521100UL
This gene is a member of the Tyr protein kinase family and the epidermal growth factor receptor subfamily It encodes a single-pass type I membrane protein with multiple cysteine rich domains a transmembrane domain a tyrosine kinase domain a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif The protein binds to and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment Mutations in this gene have been associated with cancer Alternatively spliced variants which encode different protein isoforms have been described however not all variants have been fully characterized