Learn More
Cell Signaling Technology Phospho-Erk1/2 Pathway Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502729521
Encompass
The Phospho-Erk1/2 Pathway Antibody Sampler Kit provides an economical means of evaluating multiple members of the Erk pathway as well as their activation state. The kit contains enough primary and secondary antibodies to perform two Western blot experiments.
  • Evaluates multiple members of the Erk pathway and their activation state.
  • Contains enough primary and secondary antibodies for two Western blot experiments.
  • Includes phospho-specific antibodies targeting key pathway components, such as MEK1/2, c-Raf, MSK1, p90RSK, and p44/42 MAPK (Erk1/2).
  • Validated for diverse applications including Western blotting, immunohistochemistry, and immunofluorescence.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
Catalog No. 50-272-9521 Supplier Cell Signaling Technology Supplier No. 9911T
The Phospho-Erk1/2 Pathway Antibody Sampler Kit provides an economical means of evaluating multiple members of the Erk pathway as well as their activation state. The kit contains enough primary and secondary antibodies to perform two Western blot experiments.
  • Evaluates multiple members of the Erk pathway and their activation state.
  • Contains enough primary and secondary antibodies for two Western blot experiments.
  • Includes phospho-specific antibodies targeting key pathway components, such as MEK1/2, c-Raf, MSK1, p90RSK, and p44/42 MAPK (Erk1/2).
  • Validated for diverse applications including Western blotting, immunohistochemistry, and immunofluorescence.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.