Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-ERK1 + 2 (Thr183/Tyr185) Antibody | 10 UL
SDP

Catalog No. 5000442689
Encompass_Preferred

Phospho-ERK1 + 2 (Thr183/Tyr185) Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to phospho-ERK1 + 2 (Thr183/Tyr185). It is for research use only. The antibody recognizes human and mouse species, with predicted reactivity in rat, chicken, dog, cow, horse, rabbit, and guinea pig.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M TBS (pH7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: With blue ice

Catalog No. 50-004-42689 Supplier Medchemexpress LLC Supplier No. HYP8115610UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-ERK1 + 2 (Thr183/Tyr185) Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to phospho-ERK1 + 2 (Thr183/Tyr185). It is for research use only. The antibody recognizes human and mouse species, with predicted reactivity in rat, chicken, dog, cow, horse, rabbit, and guinea pig.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M TBS (pH7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: With blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.