Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC phospho-ERK1 + 2 (Thr183/Tyr185) Antibody | 50 UL
SDP

Catalog No. 5000441374
Encompass_Preferred

phospho-ERK1 + 2 (Thr183/Tyr185) Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting phospho-ERK1 + 2 (Thr183/Tyr185). It is supplied in 0.01M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol. This antibody targets the MAP kinase family, involved in cellular processes like proliferation, differentiation, and cell cycle progression. It is activated by upstream kinases, phosphorylating nuclear targets.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, Mouse
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Applications: WB, ELISA, IHC-P, IHC-F, ICC/IF

Catalog No. 50-004-41374 Supplier Medchemexpress LLC Supplier No. HYP8115650UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

phospho-ERK1 + 2 (Thr183/Tyr185) Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting phospho-ERK1 + 2 (Thr183/Tyr185). It is supplied in 0.01M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol. This antibody targets the MAP kinase family, involved in cellular processes like proliferation, differentiation, and cell cycle progression. It is activated by upstream kinases, phosphorylating nuclear targets.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, Mouse
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Applications: WB, ELISA, IHC-P, IHC-F, ICC/IF

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.