Learn More
VAZYME INTERNATIONAL LLC VAZYME INTERNATIONAL LLC Add&Read Phospho-ERK1/2 (Thr202/Tyr204) Kit, 500 tests
Catalog No. NC3979681
Encompass
This kit is designed for the detection of phosphorylated ERK protein content in cells. It utilizes the Add&Read TR-FRET no-wash ELISA methodology, offering a streamlined two-step workflow that significantly saves time and labor compared to traditional ELISA methods.
  • Based on TR-FRET methodology (Time-Resolved Fluorescence Resonance Energy Transfer).
  • Employs a two-step workflow.
  • Saves over 80 percent of time and labor compared to traditional ELISA methods.
  • Utilizes a no-wash ELISA assay type.
  • Designed for the detection of phosphorylated ERK protein content in cells.
  • Provided in a 500-test size.
Catalog No. NC3979681 Supplier VAZYME INTERNATIONAL LLC Supplier No. DD2900102
This kit is designed for the detection of phosphorylated ERK protein content in cells. It utilizes the Add&Read TR-FRET no-wash ELISA methodology, offering a streamlined two-step workflow that significantly saves time and labor compared to traditional ELISA methods.
  • Based on TR-FRET methodology (Time-Resolved Fluorescence Resonance Energy Transfer).
  • Employs a two-step workflow.
  • Saves over 80 percent of time and labor compared to traditional ELISA methods.
  • Utilizes a no-wash ELISA assay type.
  • Designed for the detection of phosphorylated ERK protein content in cells.
  • Provided in a 500-test size.