Learn More
Cell Signaling Technology Phospho-FGF Receptor 4 Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503134052
Encompass
The Phospho-FGF Receptor 4 (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation for maximum flexibility.
  • Compatible with fluorophores, lanthanides, biotin, and beads
  • Suitable for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Formulated in 1X PBS (pH 7.8)
  • Validated for human species reactivity
  • Supplied in BSA and azide-free format
Catalog No. 50-313-4052 Supplier Cell Signaling Technology Supplier No. 19155S
The Phospho-FGF Receptor 4 (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation for maximum flexibility.
  • Compatible with fluorophores, lanthanides, biotin, and beads
  • Suitable for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Formulated in 1X PBS (pH 7.8)
  • Validated for human species reactivity
  • Supplied in BSA and azide-free format