Learn More
Abcam Phospho-gamma H2A.X (S139) ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2798161X96TESTS
Encompass_Preferred
Phospho-gamma H2A.X (S139) ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-gamma H2A.X (S139) in Human in Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2748490