Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-Glycogen synthase 1/GYS1 (S641) Antibody (YA195) | 81 kDa | 10 UL
SDP

Catalog No. 5000395672
Encompass_Preferred

This rabbit-derived, non-conjugated IgG monoclonal antibody targets Phospho-Glycogen synthase 1/GYS1 (S641) and is intended for research use only. It is suitable for applications such as Western Blotting, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, and Immunoprecipitation.

  • Recombinant, monoclonal antibody
  • Targets Phospho-Glycogen synthase 1/GYS1 (S641)
  • Species reactivity: human, mouse, rat
  • Predicted and observed molecular weight: 81 kDa
  • Protein A affinity purified
  • Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles

Catalog No. 50-003-95672 Supplier Medchemexpress LLC Supplier No. HYP8027710UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit-derived, non-conjugated IgG monoclonal antibody targets Phospho-Glycogen synthase 1/GYS1 (S641) and is intended for research use only. It is suitable for applications such as Western Blotting, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, and Immunoprecipitation.

  • Recombinant, monoclonal antibody
  • Targets Phospho-Glycogen synthase 1/GYS1 (S641)
  • Species reactivity: human, mouse, rat
  • Predicted and observed molecular weight: 81 kDa
  • Protein A affinity purified
  • Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.