Learn More
Cell Signaling Technology Phospho-HER3/ErbB3 Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131759
Encompass
The Phospho-HER3/ErbB3 (panTyr) Matched Antibody Pair is designed for high-throughput immunoassay platforms, such as ELISA, that require antibody pairs suitable for custom conjugation. The kit provides capture and detection antibodies that bind to non-overlapping epitopes on HER3/ErbB3. These antibodies are supplied in an azide- and BSA-free formulation, making them ideal for labeling with fluorophores, lanthanides, biotin, or beads for use in platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Validated for use in human samples.
  • Supplied in a concentrated, BSA and azide-free formulation in 1X PBS.
  • Compatible with diverse labeling chemistries for flexible assay development.
  • Designed for sensitive detection of phosphorylated tyrosine residues on HER3/ErbB3.
Catalog No. 50-313-1759 Supplier Cell Signaling Technology Supplier No. 85880S
The Phospho-HER3/ErbB3 (panTyr) Matched Antibody Pair is designed for high-throughput immunoassay platforms, such as ELISA, that require antibody pairs suitable for custom conjugation. The kit provides capture and detection antibodies that bind to non-overlapping epitopes on HER3/ErbB3. These antibodies are supplied in an azide- and BSA-free formulation, making them ideal for labeling with fluorophores, lanthanides, biotin, or beads for use in platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Validated for use in human samples.
  • Supplied in a concentrated, BSA and azide-free formulation in 1X PBS.
  • Compatible with diverse labeling chemistries for flexible assay development.
  • Designed for sensitive detection of phosphorylated tyrosine residues on HER3/ErbB3.