Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-HER4/ErbB4 (panTyr) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP

Catalog No. 503133660
Encompass

The Phospho-HER4/ErbB4 (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This pair is specifically optimized for applications requiring customized labeling, including conjugation with biotin, fluorophores, beads, and lanthanides. It is compatible with platforms such as Luminex, MSD, Quanterix Simoa, and Alpha Technology.

  • Contains capture and detection antibodies targeting non-overlapping epitopes.
  • Supplied in BSA and azide-free formulation for flexible conjugation.
  • Species reactivity validated in human samples.
  • Concentrated format.
  • Ideal for use in ELISA applications.

Catalog No. 50-313-3660 Supplier Cell Signaling Technology Supplier No. 53224S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-HER4/ErbB4 (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This pair is specifically optimized for applications requiring customized labeling, including conjugation with biotin, fluorophores, beads, and lanthanides. It is compatible with platforms such as Luminex, MSD, Quanterix Simoa, and Alpha Technology.

  • Contains capture and detection antibodies targeting non-overlapping epitopes.
  • Supplied in BSA and azide-free formulation for flexible conjugation.
  • Species reactivity validated in human samples.
  • Concentrated format.
  • Ideal for use in ELISA applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.