Learn More
Medchemexpress LLC Phospho-Histone H2A.X (Ser139) Antibody (YA844) | 50 UL
SDP
Catalog No. 5000443026
Encompass_Preferred
Phospho-Histone H2A.X (Ser139) Antibody (YA844) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody. This antibody targets Phospho-Histone H2A.X (Ser139) and shows reactivity with Human and Mouse samples. It is suitable for use in Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications, with an observed band size of 15 kDa. Other aliases for the target include H2A.X, H2AFX, H2a/x, HIST5-2AX, and Histone H2A.X, gamma H2A.X. The antibody is affinity purified.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets Phospho-Histone H2A.X (Ser139)
  • Reactive with human and mouse samples
  • Suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Observed band size of 15 kDa
  • Affinity purified for enhanced specificity
Catalog No. 50-004-43026 Supplier Medchemexpress LLC Supplier No. HYP8094150UL
Phospho-Histone H2A.X (Ser139) Antibody (YA844) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody. This antibody targets Phospho-Histone H2A.X (Ser139) and shows reactivity with Human and Mouse samples. It is suitable for use in Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications, with an observed band size of 15 kDa. Other aliases for the target include H2A.X, H2AFX, H2a/x, HIST5-2AX, and Histone H2A.X, gamma H2A.X. The antibody is affinity purified.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets Phospho-Histone H2A.X (Ser139)
  • Reactive with human and mouse samples
  • Suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Observed band size of 15 kDa
  • Affinity purified for enhanced specificity